missing translation for 'onlineSavingsMsg'
Learn More

9-Methylcarbazole 99.0+%, TCI America™

Catalog No. M09605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09605G 5 g
M096025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09605G Supplier TCI America Supplier No. M09605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1484-12-4
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00013431
InChI Key SDFLTYHTFPTIGX-UHFFFAOYSA-N
Synonym n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole
PubChem CID 15142
IUPAC Name 9-methylcarbazole
SMILES CN1C2=CC=CC=C2C3=CC=CC=C31

Specifications

Melting Point 89°C
Boiling Point 195°C
Quantity 5 g
Formula Weight 181.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Methylcarbazole
RTECSNumber FE6266000
TSCA No