missing translation for 'onlineSavingsMsg'
Learn More

9-Methyladenine, 98%, Thermo Scientific Chemicals

Catalog No. AC325871000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325871000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC325871000 Supplier Thermo Scientific Chemicals Supplier No. 325871000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 700-00-5 | Molecular Formula: C6H7N5 | Molecular Weight: 149.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-00-5
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00047232
InChI Key WRXCXOUDSPTXNX-UHFFFAOYSA-N
Synonym 9-methyladenine, 9-methyl-9h-purin-6-amine, n9-methyladenine, 9-methyl-9h-adenine, 6-amino-9-methylpurine, 9h-purin-6-amine, 9-methyl, adenine, 9-methyl, 9-methyl-9h-purin-6-ylamine, ccris 6905, chembl65976
PubChem CID 69689
ChEBI CHEBI:40526
IUPAC Name 9-methylpurin-6-amine
SMILES CN1C=NC2=C(N)N=CN=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300°C to 302°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 26, V,16, 128
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso
Formula Weight 149.17
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 9-Methyladenine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only