missing translation for 'onlineSavingsMsg'
Learn More

9-Methylacridine 97.0+%, TCI America™

Catalog No. p-7132610
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12371G 1 g
M12375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12371G Supplier TCI America Supplier No. M12371G

Chemical Identifiers

CAS 611-64-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00143523
InChI Key FLDRLXJNISEWNZ-UHFFFAOYSA-N
Synonym acridine, 9-methyl, 5-methylacridine, 9-methylakridin, 9-methylakridin czech, unii-1f3q4u9uis, 9-methyl acridine, 5-methylacridine european, 1f3q4u9uis, 9-methylacridin, 9-methyl-acridine
PubChem CID 11913
IUPAC Name 9-methylacridine
SMILES CC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 193.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Methylacridine
RTECSNumber AR9440000
TSCA No