missing translation for 'onlineSavingsMsg'
Learn More

9-Methyl-3,4-dihydro-2H-pyrido[1,2-a]pyrimidin-2-one 99.0+%, TCI America™

Catalog No. m067025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M067025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M067025G Supplier TCI America Supplier No. M067025G
Only null left

Chemical Identifiers

CAS 61751-44-8
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.192
MDL Number MFCD00059749
InChI Key HPVULYKIYNGGJG-UHFFFAOYSA-N
Synonym Acid Captor 9M
PubChem CID 332623
IUPAC Name 9-methyl-3,4-dihydropyrido[1,2-a]pyrimidin-2-one
SMILES CC1=CC=CN2C1=NC(=O)CC2

Specifications

Melting Point 226°C
Color White
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Methyl-3,4-dihydro-2H-pyrido[1,2-a]pyrimidin-2-one