missing translation for 'onlineSavingsMsg'
Learn More

9-Mesityl-2,7,10-trimethylacridinium Perchlorate 98.0+%, TCI America™

Catalog No. m20721g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20721G Supplier TCI America Supplier No. M20721G
Only null left

Chemical Identifiers

CAS 1216909-33-9
Molecular Formula C25H26ClNO4
Molecular Weight (g/mol) 439.94
MDL Number MFCD15072180
InChI Key VDNXOHCMBBOSPE-UHFFFAOYSA-M
PubChem CID 44629993
IUPAC Name 2,7,10-trimethyl-9-(2,4,6-trimethylphenyl)acridin-10-ium perchlorate
SMILES [O-][Cl](=O)(=O)=O.CC1=CC2=C(C3=CC(C)=CC=C3[N+](C)=C2C=C1)C1=C(C)C=C(C)C=C1C

Specifications

Quantity 1 g
UN Number 1479
Formula Weight 439.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Mesityl-2,7,10-trimethylacridinium Perchlorate
TSCA No
Recommended Storage Refrigerator