missing translation for 'onlineSavingsMsg'
Learn More

9-Hydroxyxanthene, 97+%

Catalog No. AC271520050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271520050 5 g Glass Bottle
AC271520250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271520050 Supplier Thermo Scientific Chemicals Supplier No. 271520050

CAS: 90-46-0 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-46-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00005057
InChI Key JFRMYMMIJXLMBB-UHFFFAOYSA-N
Synonym 9-hydroxyxanthene, xanthydrol, xanthanol, 9-xanthydrol, xanthen-9-ol, xanthene, hydroxy, 9-xanthenol, ccris 1640, unii-7131m69ikf, xanthrol
PubChem CID 72861
IUPAC Name 9H-xanthen-9-ol
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 124°C to 127°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Beilstein 17, 129
Solubility Information Solubility in water: insoluble
Formula Weight 198.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 9-Hydroxyxanthene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 201-996-1
RTECSNumber ZD5710000
TSCA TSCA

RUO – Research Use Only