missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Heptadecanone 98.0+%, TCI America™
$411.95 - $1403.63
Chemical Identifiers
| CAS | 540-08-9 |
|---|---|
| Molecular Formula | C17H34O |
| Molecular Weight (g/mol) | 254.458 |
| MDL Number | MFCD00009573 |
| InChI Key | WTJKUFMLQFLJOT-UHFFFAOYSA-N |
| Synonym | 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2 |
| PubChem CID | 10887 |
| IUPAC Name | heptadecan-9-one |
| SMILES | CCCCCCCCC(=O)CCCCCCCC |
Chemical Identifiers
| 540-08-9 | |
| 254.458 | |
| WTJKUFMLQFLJOT-UHFFFAOYSA-N | |
| 10887 | |
| CCCCCCCCC(=O)CCCCCCCC |
| C17H34O | |
| MFCD00009573 | |
| 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2 | |
| heptadecan-9-one |
Specifications
| 540-08-9 | |
| White-Yellow | |
| MFCD00009573 | |
| 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2 | |
| CCCCCCCCC(=O)CCCCCCCC | |
| 254.458 | |
| 254.46 | |
| Crystalline Powder |
| 54°C | |
| C17H34O | |
| 5 g | |
| WTJKUFMLQFLJOT-UHFFFAOYSA-N | |
| heptadecan-9-one | |
| 10887 | |
| ≥98.0% (GC) | |
| 9-Heptadecanone |
Safety and Handling
EINECSNumber : (2)-2475
RTECSNumber : MI3925000
TSCA : No