missing translation for 'onlineSavingsMsg'
Learn More

9-Heptadecanone, 98%

Catalog No. AAL0163014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0163014 25 g
AAL0163006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0163014 Supplier Thermo Scientific Chemicals Supplier No. L0163014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 540-08-9 | C17H34O | 254.458 g/mol

9-Heptadecanone is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Heptadecanone is used as pharmaceutical intermediates.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Avoid excessive heat and light. Always store away from incompatible compounds such as oxidizing agents.

Chemical Identifiers

CAS 540-08-9
Molecular Formula C17H34O
Molecular Weight (g/mol) 254.458
MDL Number MFCD00009573
InChI Key WTJKUFMLQFLJOT-UHFFFAOYSA-N
Synonym 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2
PubChem CID 10887
IUPAC Name heptadecan-9-one
SMILES CCCCCCCCC(=O)CCCCCCCC

Specifications

Melting Point 51°C to 53°C
Boiling Point 250°C to 253°C
Quantity 25 g
Beilstein 1707117
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 254.46
Percent Purity 98%
Chemical Name or Material 9-Heptadecanone
EINECSNumber 208-734-5
RTECSNumber MI3925000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only