missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Heptadecanone, 98%
Description
9-Heptadecanone is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Heptadecanone is used as pharmaceutical intermediates.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Avoid excessive heat and light. Always store away from incompatible compounds such as oxidizing agents.
9-Heptadecanone is used as pharmaceutical intermediates.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Avoid excessive heat and light. Always store away from incompatible compounds such as oxidizing agents.
Specifications
Specifications
| Melting Point | 51°C to 53°C |
| Boiling Point | 250°C to 253°C |
| Quantity | 5 g |
| Beilstein | 1707117 |
| Solubility Information | Soluble in toluene. Insoluble in water. |
| Formula Weight | 254.46 |
| Percent Purity | 98% |
| Chemical Name or Material | 9-Heptadecanone |
Safety and Handling
| EINECSNumber | 208-734-5 |
| RTECSNumber | MI3925000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only