missing translation for 'onlineSavingsMsg'
Learn More

9-Formyl-8-hydroxyjulolidine, 97%

CAS: 63149-33-7 | C13H15NO2 | 217.268 g/mol

$122.33 - $390.14

Chemical Identifiers

CAS 63149-33-7
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.268
MDL Number MFCD00192477
InChI Key NRZXBDYODHLZBF-UHFFFAOYSA-N
Synonym 8-hydroxy-1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline-9-carbaldehyde, 8-hydroxyjulolidine-9-aldehyde, 9-formyl-8-hydroxyjulolidine, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo ij quinolizine-9-carboxaldehyde, 8-hydroxyjulolidine-9-carboxaldehyde, 9-formyl-8-julolidinol, 1h,5h-benzo ij quinolizine-9-carboxaldehyde, 2,3,6,7-tetrahydro-8-hydroxy, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo i,j quinolizine-9-carboxaldehyde, 8-hydroxy-2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline-9-carbaldehyde
PubChem CID 113099
SMILES C1CC2=C3C(=C(C(=C2)C=O)O)CCCN3C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3178203
View Documents
Thermo Scientific Chemicals
H3178203
1 g
Each for $122.33
Only null left
 
AAH3178206
View Documents
Thermo Scientific Chemicals
H3178206
5 g
Each for $390.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63149-33-7
217.268
NRZXBDYODHLZBF-UHFFFAOYSA-N
113099
C13H15NO2
MFCD00192477
8-hydroxy-1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline-9-carbaldehyde, 8-hydroxyjulolidine-9-aldehyde, 9-formyl-8-hydroxyjulolidine, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo ij quinolizine-9-carboxaldehyde, 8-hydroxyjulolidine-9-carboxaldehyde, 9-formyl-8-julolidinol, 1h,5h-benzo ij quinolizine-9-carboxaldehyde, 2,3,6,7-tetrahydro-8-hydroxy, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo i,j quinolizine-9-carboxaldehyde, 8-hydroxy-2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline-9-carbaldehyde
C1CC2=C3C(=C(C(=C2)C=O)O)CCCN3C1

Specifications

63149-33-7
C13H15NO2
1 g
NRZXBDYODHLZBF-UHFFFAOYSA-N
217.268
217.27
9-Formyl-8-hydroxyjulolidine
72°C to 74°C
MFCD00192477
8-hydroxy-1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline-9-carbaldehyde, 8-hydroxyjulolidine-9-aldehyde, 9-formyl-8-hydroxyjulolidine, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo ij quinolizine-9-carboxaldehyde, 8-hydroxyjulolidine-9-carboxaldehyde, 9-formyl-8-julolidinol, 1h,5h-benzo ij quinolizine-9-carboxaldehyde, 2,3,6,7-tetrahydro-8-hydroxy, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo i,j quinolizine-9-carboxaldehyde, 8-hydroxy-2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline-9-carbaldehyde
C1CC2=C3C(=C(C(=C2)C=O)O)CCCN3C1
113099
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 263-966-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only