missing translation for 'onlineSavingsMsg'
Learn More

9-Ethylcarbazole 97.0+%, TCI America™

Catalog No. E007125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E007125G 25 g
E0071500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E007125G Supplier TCI America Supplier No. E007125G

Chemical Identifiers

CAS 86-28-2
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00004967
InChI Key PLAZXGNBGZYJSA-UHFFFAOYSA-N
Synonym n-ethylcarbazole, 9-ethyl-9h-carbazole, 9h-carbazole, 9-ethyl, carbazole, 9-ethyl, n-ethylcarbazol, n-ethyl carbazole, unii-6ak165l0ro, ccris 6847, 9h-ethylcarbazole, 9-ethyl carbazole
PubChem CID 6836
IUPAC Name 9-ethyl-9H-carbazole
SMILES CCN1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 68°C
Boiling Point 166°C
Quantity 25 g
Formula Weight 195.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Ethylcarbazole
EINECSNumber (5)-0151
RTECSNumber FE6225700
TSCA Yes