missing translation for 'onlineSavingsMsg'
Learn More

9-Ethylcarbazole-3-carboxaldehyde N-Ethyl-N-(m-tolyl)hydrazone 98.0+%, TCI America™

Catalog No. e05781g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05781G Supplier TCI America Supplier No. E05781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81380-24-7
Molecular Formula C24H25N3
Molecular Weight (g/mol) 355.485
MDL Number MFCD06797103
InChI Key VIRJEGJEGJEEDN-UQQQWYQISA-N
Synonym 9-Ethyl-3-[N-ethyl-N-(m-tolyl)hydrazonomethyl]carbazole
PubChem CID 93978908
IUPAC Name N-ethyl-N-[(Z)-(9-ethylcarbazol-3-yl)methylideneamino]-3-methylaniline
SMILES CCN1C2=C(C=C(C=C2)C=NN(CC)C3=CC=CC(=C3)C)C4=CC=CC=C41

Specifications

Color Yellow
Quantity 1 g
Formula Weight 355.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Ethylcarbazole-3-carboxaldehyde N-Ethyl-N-(m-tolyl)hydrazone