missing translation for 'onlineSavingsMsg'
Learn More

9-Ethylcarbazole-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E11531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11531G Supplier TCI America Supplier No. E11531G

Specifications

Chemical Name or Material 9-Ethylcarbazole-3-boronic Acid (contains varying amounts of Anhydride)
CAS 669072-93-9
Color White
Molecular Formula C14H14BNO2
Quantity 1 g
MDL Number MFCD09743121
Synonym 9-ethyl-9h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid, 9-ethyl-9h-carbazol-3-yl boronicacid
InChI Key JUBKLYCRFZTXBA-UHFFFAOYSA-N
SMILES CCN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B(O)O
IUPAC Name (9-ethyl-9H-carbazol-3-yl)boronic acid
Molecular Weight (g/mol) 239.08
PubChem CID 45382262
Formula Weight 239.08
Physical Form Crystalline Powder
Show More Show Less