missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-cis-Retinoic Acid, >98%, MP Biomedicals™
Specifications
Specifications
| CAS | 1893-1-24 |
| Assay Percent Range | ≥98% |
| Quantity | 10 mg |
| Synonym | 2,4,6-trichlorophenyl hydrazine, hydrazine, 2,4,6-trichlorophenyl, 1-2,4,6-trichlorophenyl hydrazine, 2,4,6-trichlorphenyl hydrazin, pubchem7171, acmc-1ao5o, ksc269i0b, 2,4,6 trichlorophenylhydrazine, 2,6-trichlorophenyl hydrazine |
| SMILES | C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O |
| IUPAC Name | (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid |
| PubChem CID | 79234 |
| Formula Weight | 300.44 |
| Physical Form | Powder |
| Chemical Name or Material | 9-cis-Retinoic Acid, Cell Biology and Analysis |
Safety and Handling
| Recommended Storage | Store at -20°C. |