missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(Bromomethyl)acridine 98.0+%, TCI America™
$285.21 - $784.76
Chemical Identifiers
| CAS | 1556-34-9 |
|---|---|
| Molecular Formula | C14H10BrN |
| Molecular Weight (g/mol) | 272.15 |
| MDL Number | MFCD01321146 |
| InChI Key | MZFYKBHQWLWIBI-UHFFFAOYSA-N |
| PubChem CID | 150930 |
| IUPAC Name | 9-(bromomethyl)acridine |
| SMILES | BrCC1=C2C=CC=CC2=NC2=CC=CC=C12 |
Chemical Identifiers
| 1556-34-9 | |
| 272.15 | |
| MZFYKBHQWLWIBI-UHFFFAOYSA-N | |
| 9-(bromomethyl)acridine |
| C14H10BrN | |
| MFCD01321146 | |
| 150930 | |
| BrCC1=C2C=CC=CC2=NC2=CC=CC=C12 |
Specifications
| 1556-34-9 | |
| C14H10BrN | |
| 1 g | |
| BrCC1=C2C=CC=CC2=NC2=CC=CC=C12 | |
| 272.15 | |
| 272.15 | |
| Crystalline Powder |
| 165°C | |
| MFCD01321146 | |
| MZFYKBHQWLWIBI-UHFFFAOYSA-N | |
| 9-(bromomethyl)acridine | |
| 150930 | |
| ≥98.0% (HPLC,T) | |
| 9-(Bromomethyl)acridine [for HPLC Labeling] |
Safety and Handling
TSCA : No