missing translation for 'onlineSavingsMsg'
Learn More

9-(Bromomethyl)acridine 98.0+%, TCI America™

Catalog No. B19261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19261G 1 g
B19265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19261G Supplier TCI America Supplier No. B19261G
Only null left

Chemical Identifiers

CAS 1556-34-9
Molecular Formula C14H10BrN
Molecular Weight (g/mol) 272.15
MDL Number MFCD01321146
InChI Key MZFYKBHQWLWIBI-UHFFFAOYSA-N
PubChem CID 150930
IUPAC Name 9-(bromomethyl)acridine
SMILES BrCC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Melting Point 165°C
Quantity 1 g
Formula Weight 272.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-(Bromomethyl)acridine [for HPLC Labeling]