missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-phenylanthracene 98.0+%, TCI America™

Catalog No. B39771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39771G 1 g
B39775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39771G Supplier TCI America Supplier No. B39771G

Chemical Identifiers

CAS 23674-20-6
Molecular Formula C20H13Br
Molecular Weight (g/mol) 333.228
MDL Number MFCD00230983
InChI Key WHGGVVHVBFMGSG-UHFFFAOYSA-N
Synonym 10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene
PubChem CID 4155836
IUPAC Name 9-bromo-10-phenylanthracene
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br

Specifications

Melting Point 156°C
Color Yellow
Quantity 1 g
Formula Weight 333.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-phenylanthracene