missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-(2-naphthyl)anthracene 98.0+%, TCI America™

Catalog No. p-7123529
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44261G 1 g
B44265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44261G Supplier TCI America Supplier No. B44261G
Only null left

Chemical Identifiers

CAS 474688-73-8
Molecular Formula C24H15Br
Molecular Weight (g/mol) 383.288
MDL Number MFCD09832882
InChI Key FKIFDWYMWOJKTQ-UHFFFAOYSA-N
Synonym 9-bromo-10-2-naphthyl anthracene, 9-bromo-10-naphthalen-2-yl anthracene, 10-bromo-9-naphthalen-2-yl anthracene, anthracene, 9-bromo-10-2-naphthalenyl, ksc235k9l, 10-2-naphthyl-9-anthryl bromide, 9-bromo-10-2-naphthyl-anthracene, 10-bromo-9-phthalen-2-yl anthracene, 9-bromo-10-naphthyl-2-yl anthracene
PubChem CID 16116263
IUPAC Name 9-bromo-10-naphthalen-2-ylanthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br

Specifications

Melting Point 171°C
Color Yellow
Quantity 1 g
Formula Weight 383.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-(2-naphthyl)anthracene