missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-(1-naphthyl)anthracene 98.0+%, TCI America™

Catalog No. B44515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44515G 5 g
B44511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44515G Supplier TCI America Supplier No. B44515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 400607-04-7
Molecular Formula C24H15Br
Molecular Weight (g/mol) 383.288
MDL Number MFCD11046571
InChI Key SYACRXBYRNYMLN-UHFFFAOYSA-N
Synonym 9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
PubChem CID 21076365
IUPAC Name 9-bromo-10-naphthalen-1-ylanthracene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br

Specifications

Melting Point 179°C
Color Yellow
Quantity 5 g
Formula Weight 383.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-(1-naphthyl)anthracene