missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-(1,1':3',1″-terphenyl-5'-yl)anthracene 98.0+%, TCI America™

Catalog No. B4755200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4755200MG 200 mg
B47551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4755200MG Supplier TCI America Supplier No. B4755200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 474688-74-9
Molecular Formula C32H21Br
Molecular Weight (g/mol) 485.424
MDL Number MFCD26406220
InChI Key BQYDXENQXPPQLP-UHFFFAOYSA-N
PubChem CID 23067325
IUPAC Name 9-bromo-10-(3,5-diphenylphenyl)anthracene
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br)C6=CC=CC=C6

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 485.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-(1,1′:3′,1′′-terphenyl-5′-yl)anthracene