missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-1-nonanol 95.0+%, TCI America™

Catalog No. p-7119261
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17855G 5 g
B178525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17855G Supplier TCI America Supplier No. B17855G
Only null left

Chemical Identifiers

CAS 55362-80-6
Molecular Formula C9H19BrO
Molecular Weight (g/mol) 223.15
MDL Number MFCD00063348
InChI Key USJDOLXCPFASNV-UHFFFAOYSA-N
Synonym 9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq
PubChem CID 108700
IUPAC Name 9-bromononan-1-ol
SMILES OCCCCCCCCCBr

Specifications

Boiling Point 126°C
Quantity 5 g
Formula Weight 223.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 9-Bromo-1-nonanol
TSCA Yes
Recommended Storage Refrigerator