missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-1-nonanol, 98%

Catalog No. AAH6139314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6139314 25 g
AAH6139306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6139314 Supplier Thermo Scientific Chemicals Supplier No. H6139314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55362-80-6 | C9H19BrO | 223.15 g/mol

9-Bromo-1-nonanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromo-1-nonanol is used as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, ethyl acetate and hexane.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 55362-80-6
Molecular Formula C9H19BrO
Molecular Weight (g/mol) 223.15
MDL Number MFCD00063348
InChI Key USJDOLXCPFASNV-UHFFFAOYSA-N
Synonym 9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq
PubChem CID 108700
IUPAC Name 9-bromononan-1-ol
SMILES OCCCCCCCCCBr

Specifications

Melting Point 33°C to 35°C
Boiling Point 125°C to 126°C (2 mmHg)
Flash Point 109°C (228°F)
Linear Formula Br(CH2)9OH
Quantity 25 g
Beilstein 1737525
Solubility Information Soluble in dichloromethane,ethyl acetate and hexane.
Formula Weight 223.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-Bromo-1-nonanol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only