missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosylguanine Hydrate 98.0+%, TCI America™

Catalog No. p-7118049
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
A273910MG 10 mg
A273950MG 50 mg
2 options

Catalog No. A273910MG

Supplier: TCI America A273910MG

Only null left

Chemical Identifiers

CAS 38819-10-2
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00065486
InChI Key NYHBQMYGNKIUIF-FJFJXFQQSA-N
Synonym Ara-G, Guanine 9-beta-D-Arabinofuranoside
PubChem CID 65412
ChEBI CHEBI:73141
IUPAC Name 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 225°C
Color White
Quantity 10 mg
Formula Weight 283.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-beta-D-Arabinofuranosylguanine Hydrate
RTECSNumber MF8301000
TSCA No
Recommended Storage Refrigerator