missing translation for 'onlineSavingsMsg'
Learn More

9-Benzyl-3-bromo-9H-carbazole 98.0+%, TCI America™

Catalog No. b54611g
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B54611G 1 g
B54615G 5 g
2 options

Catalog No. B54611G

Supplier: TCI America B54611G

Only null left

Chemical Identifiers

CAS 339576-55-5
Molecular Formula C19H14BrN
Molecular Weight (g/mol) 336.232
MDL Number MFCD00821698
InChI Key NMNQVRIUUBFICK-UHFFFAOYSA-N
PubChem CID 4379666
IUPAC Name 9-benzyl-3-bromocarbazole
SMILES C1=CC=C(C=C1)CN2C3=C(C=C(C=C3)Br)C4=CC=CC=C42

Specifications

Melting Point 118°C
Quantity 1 g
Formula Weight 336.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzyl-3-bromo-9H-carbazole