missing translation for 'onlineSavingsMsg'
Learn More

9-Benzyl-2,7-dibromo-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7124088
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54621G 1 g
B54625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54621G Supplier TCI America Supplier No. B54621G

Chemical Identifiers

CAS 1384281-49-5
Molecular Formula C19H13Br2N
Molecular Weight (g/mol) 415.128
InChI Key HJQQHWYCZFAJHY-UHFFFAOYSA-N
PubChem CID 50936718
IUPAC Name 9-benzyl-2,7-dibromocarbazole
SMILES C1=CC=C(C=C1)CN2C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 1 g
Formula Weight 415.13
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzyl-2,7-dibromo-9H-carbazole