missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoacridine Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. A06195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06195G 5 g
A061925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06195G Supplier TCI America Supplier No. A06195G
Only null left

Chemical Identifiers

CAS 52417-22-8
Molecular Formula C13H11ClN2
Molecular Weight (g/mol) 230.70
MDL Number MFCD00012663
InChI Key FTGPOQQGJVJDCT-UHFFFAOYSA-N
Synonym 9-aminoacridine hydrochloride monohydrate, acridin-9-amine hydrochloride hydrate, 9-aminoacridine hydrochloride hydrate, 9-acridinamine, monohydrochloride, monohydrate, 9-aminoacridine, monohydrochloride, monohydrate, acridin-9-ylamine, dsstox_cid_4457, dsstox_rid_77405, dsstox_gsid_24457, acridine-9-ylamine, chloride, hydrate
PubChem CID 2723598
IUPAC Name acridin-9-amine hydrochloride
SMILES Cl.NC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Color Yellow
Quantity 5 g
Formula Weight 230.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Aminoacridine Hydrochloride Monohydrate
RTECSNumber AR6850000
TSCA Yes