missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Dimethylxanthene 98.0+%, TCI America™
$59.65 - $167.76
Chemical Identifiers
| CAS | 19814-75-6 |
|---|---|
| Molecular Formula | C15H14O |
| Molecular Weight (g/mol) | 210.28 |
| MDL Number | MFCD00134434 |
| InChI Key | MTVNAPYHLASOSX-UHFFFAOYSA-N |
| Synonym | 9,9-dimethyl-9h-xanthene, 9h-xanthene, 9,9-dimethyl, 9,9-dimethylxantene, 9,9-dimethyl xanthene, xanthene, 9,9-dimethyl, ksc182m0n, 9,9-dimethylxanthene, 9,9-dimethyl-9h-xanthene # |
| PubChem CID | 606997 |
| IUPAC Name | 9,9-dimethyl-9H-xanthene |
| SMILES | CC1(C)C2=CC=CC=C2OC2=CC=CC=C12 |
Chemical Identifiers
| 19814-75-6 | |
| 210.28 | |
| MTVNAPYHLASOSX-UHFFFAOYSA-N | |
| 606997 | |
| CC1(C)C2=CC=CC=C2OC2=CC=CC=C12 |
| C15H14O | |
| MFCD00134434 | |
| 9,9-dimethyl-9h-xanthene, 9h-xanthene, 9,9-dimethyl, 9,9-dimethylxantene, 9,9-dimethyl xanthene, xanthene, 9,9-dimethyl, ksc182m0n, 9,9-dimethylxanthene, 9,9-dimethyl-9h-xanthene # | |
| 9,9-dimethyl-9H-xanthene |
Specifications
| 19814-75-6 | |
| White-Yellow | |
| C15H14O | |
| 1 g | |
| MTVNAPYHLASOSX-UHFFFAOYSA-N | |
| 9,9-dimethyl-9H-xanthene | |
| 606997 | |
| ≥98.0% (GC) | |
| 9,9-Dimethylxanthene |
| 38°C | |
| 115°C | |
| MFCD00134434 | |
| 9,9-dimethyl-9h-xanthene, 9h-xanthene, 9,9-dimethyl, 9,9-dimethylxantene, 9,9-dimethyl xanthene, xanthene, 9,9-dimethyl, ksc182m0n, 9,9-dimethylxanthene, 9,9-dimethyl-9h-xanthene # | |
| CC1(C)C2=CC=CC=C2OC2=CC=CC=C12 | |
| 210.28 | |
| 210.28 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No