missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethyl-9H-9-silafluorene 98.0+%, TCI America™

Catalog No. D43081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43081G 1 g
D43085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43081G Supplier TCI America Supplier No. D43081G

Chemical Identifiers

CAS 13688-68-1
Molecular Formula C14H14Si
Molecular Weight (g/mol) 210.351
InChI Key YJQFTKVGILQPMC-UHFFFAOYSA-N
Synonym 5,5-Dimethyl-5H-dibenzo[b,d]silole
PubChem CID 139544
IUPAC Name 5,5-dimethylbenzo[b][1]benzosilole
SMILES C[Si]1(C2=CC=CC=C2C3=CC=CC=C31)C

Specifications

Melting Point 60°C
Color White
Quantity 1 g
Formula Weight 210.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Dimethyl-9H-9-silafluorene