missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific™

Catalog No. p-7038067
Change view
Click to view available options
Quantity:
25g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5695214 25g
AAH5695222 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5695214 Supplier Thermo Scientific Chemicals Supplier No. H5695214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 189367-54-2
Molecular Formula C25H32Br2
Molecular Weight (g/mol) 492.339
MDL Number MFCD03427215
InChI Key OXFFIMLCSVJMHA-UHFFFAOYSA-N
Synonym 9,9-dihexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene, 2,7-dibromo-9,9-di-n-hexylfluorene, 9h-fluorene, 2,7-dibromo-9,9-dihexyl, 2,7-dibromo-9,9-dihexyl-fluorene, acmc-1c0sw, ksc491i1r, 9,9-di-n-hexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene homopolymer
PubChem CID 3539647
IUPAC Name 2,7-dibromo-9,9-dihexylfluorene
SMILES CCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCC

Specifications

Quantity 25g
Solubility Information Slightly soluble in water.
Formula Weight 492.33
Percent Purity 98%
Chemical Name or Material 9,9-Di-n-hexyl-2,7-dibromofluorene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only