missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis(4-glycidyloxyphenyl)fluorene 98.0+%, TCI America™
$108.11 - $320.62
Chemical Identifiers
| CAS | 47758-37-2 |
|---|---|
| Molecular Formula | C31H26O4 |
| Molecular Weight (g/mol) | 462.55 |
| MDL Number | MFCD02171148 |
| InChI Key | LCSAOPVSVLGDLE-UHFFFAOYNA-N |
| PubChem CID | 93249 |
| IUPAC Name | 2-{[4-(9-{4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane |
| SMILES | C(OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC2CO2)C=C1)C1CO1 |
Chemical Identifiers
| 47758-37-2 | |
| 462.55 | |
| LCSAOPVSVLGDLE-UHFFFAOYNA-N | |
| 2-{[4-(9-{4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane |
| C31H26O4 | |
| MFCD02171148 | |
| 93249 | |
| C(OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC2CO2)C=C1)C1CO1 |
Specifications
| 47758-37-2 | |
| White | |
| MFCD02171148 | |
| 4,4′C-(9-Fluorenylidene)bis(1,2-epoxy-3-phenoxypropane) | |
| C(OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC2CO2)C=C1)C1CO1 | |
| 462.55 | |
| 462.55 | |
| Crystalline Powder |
| 159°C | |
| C31H26O4 | |
| 5 g | |
| LCSAOPVSVLGDLE-UHFFFAOYNA-N | |
| 2-{[4-(9-{4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane | |
| 93249 | |
| ≥98.0% (GC) | |
| 9,9-Bis(4-glycidyloxyphenyl)fluorene |
Safety and Handling
EINECSNumber : (4)-1713
TSCA : Yes