missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Bianthracene 98.0+%, TCI America™

Catalog No. B40955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40955G 5 g
B40951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40955G Supplier TCI America Supplier No. B40955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1055-23-8
Molecular Formula C28H18
Molecular Weight (g/mol) 354.45
MDL Number MFCD00130216
InChI Key SXGIRTCIFPJUEQ-UHFFFAOYSA-N
Synonym 9,9'-bianthracene, 9,9'-bianthryl, 9,9'-bianthracenyl, 9-9-anthryl anthracene, 9,9'-bianthranyl, 9,9'-dianthracene, 9,9-bianthracene, 9,9/'-bianthracene, 9,9 inverted exclamation mark-bianthracene, 9-9-anthracenyl anthracene
PubChem CID 66105
IUPAC Name 9,9'-bianthracene
SMILES C1=CC2=CC3=CC=CC=C3C(=C2C=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 306°C
Quantity 5 g
Formula Weight 354.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′-Bianthracene
RTECSNumber DT3572500
TSCA No