missing translation for 'onlineSavingsMsg'
Learn More

9,9'-(5-Bromo-1,3-phenylene)bis(9H-carbazole) 98.0+%, TCI America™

Catalog No. p-7124101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55021G 1 g
B55025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55021G Supplier TCI America Supplier No. B55021G

Chemical Identifiers

CAS 750573-24-1
Molecular Formula C30H19BrN2
Molecular Weight (g/mol) 487.40
MDL Number MFCD28100481
InChI Key SJOKONNBSXFPSN-UHFFFAOYSA-N
Synonym 1-Bromo-3,5-bis(carbazol-9-yl)benzene
PubChem CID 59770826
IUPAC Name 9-[3-bromo-5-(9H-carbazol-9-yl)phenyl]-9H-carbazole
SMILES BrC1=CC(=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 1 g
Formula Weight 487.40
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′-(5-Bromo-1,3-phenylene)bis(9H-carbazole)