missing translation for 'onlineSavingsMsg'
Learn More

9,9',10,10'-Tetraphenyl-2,2'-bianthracene 97.0+%, TCI America™

Catalog No. p-7137703
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3336200MG 200 mg
T33361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3336200MG Supplier TCI America Supplier No. T3336200MG
Only null left

Chemical Identifiers

CAS 172285-72-2
Molecular Formula C52H34
Molecular Weight (g/mol) 658.84
MDL Number MFCD12022455
InChI Key BHPFDLWDNJSMOS-UHFFFAOYSA-N
PubChem CID 23154896
IUPAC Name 9,9',10,10'-tetraphenyl-2,2'-bianthracene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC(=CC=C12)C1=CC=C2C(=C1)C(C1=CC=CC=C1)=C1C=CC=CC1=C2C1=CC=CC=C1

Specifications

Quantity 200 mg
Formula Weight 658.84
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′,10,10′-Tetraphenyl-2,2′-bianthracene