missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(6-Bromohexyl)-9H-carbazole 97.0+%, TCI America™
$138.61 - $411.98
Chemical Identifiers
| CAS | 94847-10-6 |
|---|---|
| Molecular Formula | C18H20BrN |
| Molecular Weight (g/mol) | 330.27 |
| MDL Number | MFCD30562129 |
| InChI Key | HFNNZEMVTJAKRC-UHFFFAOYSA-N |
| PubChem CID | 11738992 |
| IUPAC Name | 9-(6-bromohexyl)-9H-carbazole |
| SMILES | BrCCCCCCN1C2=C(C=CC=C2)C2=C1C=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5548200MG
|
TCI America
B5548200MG |
200 mg |
Each for $138.61
|
|
|||||
|
B55481G
|
TCI America
B55481G |
1 g |
Each for $411.98
|
|
|||||
Chemical Identifiers
| 94847-10-6 | |
| 330.27 | |
| HFNNZEMVTJAKRC-UHFFFAOYSA-N | |
| 9-(6-bromohexyl)-9H-carbazole |
| C18H20BrN | |
| MFCD30562129 | |
| 11738992 | |
| BrCCCCCCN1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Specifications
| 94847-10-6 | |
| White-Yellow | |
| MFCD30562129 | |
| HFNNZEMVTJAKRC-UHFFFAOYSA-N | |
| 9-(6-bromohexyl)-9H-carbazole | |
| 11738992 | |
| ≥97.0% (GC) | |
| 9-(6-Bromohexyl)-9H-carbazole |
| 60°C | |
| C18H20BrN | |
| 200 mg | |
| BrCCCCCCN1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
| 330.27 | |
| 330.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No