missing translation for 'onlineSavingsMsg'
Learn More

9-[4-(tert-Butyl)phenyl]-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7124144
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56201G 1 g
B56205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56201G Supplier TCI America Supplier No. B56201G
Only null left

Chemical Identifiers

CAS 57103-13-6
Molecular Formula C18H25N5O
Molecular Weight (g/mol) 327.432
InChI Key IPFLSUSUYBRVNV-UHFFFAOYSA-N
PubChem CID 44301296
IUPAC Name 5-[(8-methoxy-2,2,4-trimethyl-3,4-dihydro-1H-quinolin-6-yl)methyl]pyrimidine-2,4-diamine
SMILES CC1CC(NC2=C(C=C(C=C12)CC3=CN=C(N=C3N)N)OC)(C)C

Specifications

Melting Point 191°C
Color White
Quantity 1 g
Formula Weight 299.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-[4-(tert-Butyl)phenyl]-9H-carbazole