missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(4-Ethynylphenyl)carbazole 98.0+%, TCI America™
Supplier: TCI America E10551G
Specifications
| 9-(4-Ethynylphenyl)carbazole | |
| 106°C | |
| C20H13N | |
| 1 g | |
| C#CC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
| 267.33 | |
| 267.33 | |
| Crystalline Powder |
| 262861-81-4 | |
| White-Yellow | |
| MFCD12024288 | |
| JJIQKIUIIVFHAN-UHFFFAOYSA-N | |
| 9-(4-ethynylphenyl)-9H-carbazole | |
| 15606338 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 262861-81-4 | |
| 267.33 | |
| JJIQKIUIIVFHAN-UHFFFAOYSA-N | |
| 9-(4-ethynylphenyl)-9H-carbazole |
| C20H13N | |
| MFCD12024288 | |
| 15606338 | |
| C#CC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Safety and Handling
TSCA : No