missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Bromophenyl)carbazole 98.0+%, TCI America™

Catalog No. B35541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35541G 1 g
B35545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35541G Supplier TCI America Supplier No. B35541G

Chemical Identifiers

CAS 57102-42-8
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.205
MDL Number MFCD11617969
InChI Key XSDKKRKTDZMKCH-UHFFFAOYSA-N
Synonym 9-4-bromophenyl-9h-carbazole, 9-4-bromophenyl carbazole, 9-4-bromo-phenyl-9h-carbazole, 9h-carbazole, 9-4-bromophenyl, acmc-209lw0, n-4-bromophenyl carbazole, amth018
PubChem CID 22361390
IUPAC Name 9-(4-bromophenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)Br

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 1 g
Formula Weight 322.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Bromophenyl)carbazole