missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Bromophenyl)carbazole, 98%, Thermo Scientific Chemicals

Catalog No. p-7055748
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6440403 1 g
AAH6440406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6440403 Supplier Thermo Scientific Chemicals Supplier No. H6440403

It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 57102-42-8
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.205
MDL Number MFCD11617969
InChI Key XSDKKRKTDZMKCH-UHFFFAOYSA-N
Synonym 9-4-bromophenyl-9h-carbazole, 9-4-bromophenyl carbazole, 9-4-bromo-phenyl-9h-carbazole, 9h-carbazole, 9-4-bromophenyl, acmc-209lw0, n-4-bromophenyl carbazole, amth018
PubChem CID 22361390
IUPAC Name 9-(4-bromophenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)Br

Specifications

Melting Point 147°C to 151°C
Quantity 1 g
Beilstein 199590
Solubility Information Slightly soluble in water.
Formula Weight 322.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-(4-Bromophenyl)carbazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only