missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Bromophenyl)-10-phenylanthracene 98.0+%, TCI America™

Catalog No. b4475200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4475200MG 200 mg
B44751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4475200MG Supplier TCI America Supplier No. B4475200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625854-02-6
Molecular Formula C26H17Br
Molecular Weight (g/mol) 409.326
MDL Number MFCD24842536
InChI Key LDFCHUHQZQRSHF-UHFFFAOYSA-N
PubChem CID 22138275
IUPAC Name 9-(4-bromophenyl)-10-phenylanthracene
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)C5=CC=C(C=C5)Br

Specifications

Melting Point 227°C
Quantity 200 mg
Formula Weight 409.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Bromophenyl)-10-phenylanthracene