missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(4-Biphenylyl)-3-bromocarbazole 98.0+%, TCI America™
Supplier: TCI America B453825G
Specifications
| 9-(4-Biphenylyl)-3-bromocarbazole | |
| 155°C | |
| C24H16BrN | |
| 25 g | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 | |
| 398.303 | |
| 398.30 | |
| Crystalline Powder |
| 894791-46-9 | |
| White-Yellow | |
| MFCD16038226 | |
| MOCNGNGLTRMQQH-UHFFFAOYSA-N | |
| 3-bromo-9-(4-phenylphenyl)carbazole | |
| 53401105 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 894791-46-9 | |
| 398.303 | |
| MOCNGNGLTRMQQH-UHFFFAOYSA-N | |
| 3-bromo-9-(4-phenylphenyl)carbazole |
| C24H16BrN | |
| MFCD16038226 | |
| 53401105 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Safety and Handling
TSCA : No