missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Biphenylyl)-3-(4-bromophenyl)carbazole 98.0+%, TCI America™

Catalog No. p-7123962
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5111200MG 200 mg
B51111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5111200MG Supplier TCI America Supplier No. B5111200MG

Chemical Identifiers

CAS 1028648-25-0
Molecular Formula C30H20BrN
Molecular Weight (g/mol) 474.401
MDL Number MFCD22571689
InChI Key AZGVZMFTHCOVID-UHFFFAOYSA-N
Synonym 9-([1,1′C-Biphenyl]-4-yl)-3-(4-bromophenyl)-9H-carbazole
PubChem CID 59728582
IUPAC Name 3-(4-bromophenyl)-9-(4-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)C5=CC=C(C=C5)Br)C6=CC=CC=C63

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 200 mg
Formula Weight 474.40
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Biphenylyl)-3-(4-bromophenyl)carbazole