missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Biphenylyl)-2-bromocarbazole 98.0+%, TCI America™

Catalog No. B5110200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5110200MG 200 mg
B51101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5110200MG Supplier TCI America Supplier No. B5110200MG

Chemical Identifiers

CAS 1393835-87-4
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.303
MDL Number MFCD27979953
InChI Key WNJONVVVRMCDDZ-UHFFFAOYSA-N
Synonym 9-([1,1′C-Biphenyl]-4-yl)-2-bromo-9H-carbazole
PubChem CID 68191373
IUPAC Name 2-bromo-9-(4-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 200 mg
Formula Weight 398.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Biphenylyl)-2-bromocarbazole