missing translation for 'onlineSavingsMsg'
Learn More

9-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole 98.0+%, TCI America™

Catalog No. T34525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34525G 5 g
T34521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34525G Supplier TCI America Supplier No. T34525G

Chemical Identifiers

CAS 785051-54-9
Molecular Formula C24H24BNO2
Molecular Weight (g/mol) 369.271
MDL Number MFCD16294549
InChI Key AHDSYMVAUJZCOP-UHFFFAOYSA-N
Synonym 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbazole, amtb250, 9-carbazolylbenzeneboronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid pinacol ester, 9h-carbazole-9-4-phenyl boronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid, pinacol ester
PubChem CID 11850073
IUPAC Name 9-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 5 g
Formula Weight 369.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole