missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(3-Biphenylyl)-3-bromocarbazole 98.0+%, TCI America™
$72.70 - $361.62
Chemical Identifiers
| CAS | 1428551-28-3 |
|---|---|
| Molecular Formula | C24H16BrN |
| Molecular Weight (g/mol) | 398.303 |
| MDL Number | MFCD28138081 |
| InChI Key | NSRPRPVECXNOLB-UHFFFAOYSA-N |
| PubChem CID | 89472056 |
| IUPAC Name | 3-bromo-9-(3-phenylphenyl)carbazole |
| SMILES | C1=CC=C(C=C1)C2=CC(=CC=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Chemical Identifiers
| 1428551-28-3 | |
| 398.303 | |
| NSRPRPVECXNOLB-UHFFFAOYSA-N | |
| 3-bromo-9-(3-phenylphenyl)carbazole |
| C24H16BrN | |
| MFCD28138081 | |
| 89472056 | |
| C1=CC=C(C=C1)C2=CC(=CC=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Specifications
| 1428551-28-3 | |
| White-Yellow | |
| MFCD28138081 | |
| NSRPRPVECXNOLB-UHFFFAOYSA-N | |
| 3-bromo-9-(3-phenylphenyl)carbazole | |
| 89472056 | |
| ≥98.0% (GC) | |
| 9-(3-Biphenylyl)-3-bromocarbazole |
| 110°C | |
| C24H16BrN | |
| 1 g | |
| C1=CC=C(C=C1)C2=CC(=CC=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 | |
| 398.303 | |
| 398.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No