missing translation for 'onlineSavingsMsg'
Learn More

9-(3-Biphenylyl)-3-bromocarbazole 98.0+%, TCI America™

Catalog No. B50245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50245G 5 g
B50241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50245G Supplier TCI America Supplier No. B50245G
Only null left

Chemical Identifiers

CAS 1428551-28-3
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.303
MDL Number MFCD28138081
InChI Key NSRPRPVECXNOLB-UHFFFAOYSA-N
PubChem CID 89472056
IUPAC Name 3-bromo-9-(3-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 5 g
Formula Weight 398.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(3-Biphenylyl)-3-bromocarbazole