missing translation for 'onlineSavingsMsg'
Learn More

9,3':6',9″-Tercarbazole 98.0+%, TCI America™

Catalog No. T3344200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3344200MG 200 mg
T33441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3344200MG Supplier TCI America Supplier No. T3344200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606129-90-2
Molecular Formula C36H23N3
Molecular Weight (g/mol) 497.601
InChI Key OGDZAJUZGODBKX-UHFFFAOYSA-N
PubChem CID 12103890
IUPAC Name 3,6-di(carbazol-9-yl)-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC5=C(C=C4)NC6=C5C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97

Specifications

Melting Point 285°C
Quantity 200 mg
Formula Weight 497.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,3′:6′,9′′-Tercarbazole