missing translation for 'onlineSavingsMsg'
Learn More

9-(2-Naphthyl)anthracene 98.0+%, TCI America™

Catalog No. N11301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11301G 1 g
N11305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N11301G Supplier TCI America Supplier No. N11301G

Chemical Identifiers

CAS 7424-72-8
Molecular Formula C24H16
Molecular Weight (g/mol) 304.392
MDL Number MFCD18205835
InChI Key MFDORGWIGJJZEQ-UHFFFAOYSA-N
PubChem CID 264416
IUPAC Name 9-naphthalen-2-ylanthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=CC5=CC=CC=C53

Specifications

Melting Point 205°C
Quantity 1 g
Formula Weight 304.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(2-Naphthyl)anthracene