missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 405.39 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Starch Syrup |
| Chemical Name or Material | 9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole |
Safety and Handling
| TSCA | No |