missing translation for 'onlineSavingsMsg'
Learn More

9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™

Catalog No. E1323200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1323200MG 200 mg
E13231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1323200MG Supplier TCI America Supplier No. E1323200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 856422-41-8
Molecular Formula C26H36BNO2
Synonym 9-(2-Ethylhexyl)-9H-carbazole-3-boronic Acid Pinacol Ester, 2-[9-(2-Ethylhexyl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 405.39
Percent Purity ≥98.0% (HPLC)
Physical Form Starch Syrup at 20°C
Chemical Name or Material 9-(2-Ethylhexyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole