missing translation for 'onlineSavingsMsg'
Learn More

9-(2-Bromophenyl)-9H-carbazole 98.0+%, TCI America™

Catalog No. B41091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41091G 1 g
B41095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41091G Supplier TCI America Supplier No. B41091G
Only null left

Chemical Identifiers

CAS 902518-11-0
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.21
MDL Number MFCD23135883
InChI Key KEWDVYIULXXMPP-UHFFFAOYSA-N
PubChem CID 59465012
IUPAC Name 9-(2-bromophenyl)-9H-carbazole
SMILES BrC1=CC=CC=C1N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 1 g
Formula Weight 322.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(2-Bromophenyl)-9H-carbazole